Laboratory trays
Supplier: KARTELL
548
216-1466EA 48.7 EUR
216-1466
Laboratory trays
Trays/Dishes/Pots Laboratory Trays/Dishes
PP, translucent.
  • Excellent chemical and thermal resistance
  • Particularly deep design for increased stability
  • Versatile e.g. for transporting and storing laboratory utensils