Single wafer shippers, H 22 series
Supplier: ENTEGRIS
H22-101-0615 H22-20-0615 H22-10-0615 H22-151-0615 H22-15-0615 H22-102-0815 H22-251-0615 H22-252-0615 H22-201-0615 H22-102-0615 H22-25-0615_U H22-152-0615 H22-202A-0615
216-1578EA 0 EUR
216-1578 216-1582 216-1577 216-1609 216-1580 216-2067 216-1586 216-1587 216-1583 216-1579 ENTEH22-25-0615_U 216-1581 216-2074
Single wafer shippers, H 22 series
Wafer Handling Supplies Wafer Shippers
Available for 1,0"; 1,5"; 2,0" and 2,5" wafers. Large area on cover provides space for labelling.
  • Conical pocket holds one wafer face down contacting only the edge of the wafer
  • Spring retains wafer and minimises particle generation
  • Stackable for efficient use of storage space
  • Loose or lock stacking, either tray on tray or cover on tray
  • Convenient screw on covers
Please contact Avantor for complete range of products and options.
Caution: Articles may not be available in all countries. Please contact your local Avantor office.