VWR® Transport and Storage Containers, Multi-Use Box
Supplier: VWR Collection
216-2378EA 52.1 EUR
216-2378 216-2379 216-2380 216-2381 216-2383 216-2382
VWR® Transport and Storage Containers, Multi-Use Box
Boxes Transport Boxes
Made of transparent virgin polypropylene which is food grade. Multi-use boxes are strong, transparent plastic containers that can be used to pack, store and transport a wide variety of products, including foodstuffs.
  • Transparent colour makes the contents easily visible
  • Hygienic smooth walls for easy cleaning
  • Food grade, freezer and dishwasher safe
  • Suitable for –25 to +95 °C storage
  • Stackable when lid is in place, nestable without the lids when not in use
  • 90% space saving when empty
  • Lid is secured in place with the clip handles
Available in large,More Product Information