Bowl, hemispherical mould
Supplier: CONTACTO
877/200
216-7425EA 0 EUR
216-7425
Bowl, hemispherical mould
Trays/Dishes/Pots Laboratory Trays/Dishes
Stainless steel 18/10.
  • Mirror polished finish
  • Extra heavy gauge
  • Good chemical resistance, very good thermal resistance
Multipurpose bowl for a wide range of uses.