Barium carbonate ≥99.0%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Warning

21702.235EA 85 EUR
21702.235 21702.292
Barium carbonate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Barium carbonate
Formula: BaCO₃
Melting Pt: 1350 °C
Density: 3.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1,III
Merck Index: 13,00972

Order Now

Specification Test Results

Assay Min. 99.0 %
Alkali hydroxides+carbonates(as Ba(OH)2) Max. 0.015 %
Heavy metals (as Pb) Max. 10 ppm
Insolubility in hydrochloric acid Max. 0.015 %
Oxidising substances (as NO3) Max. 50 ppm
S (Sulphur) Max. 10 ppm
Ca (Calcium) Max. 0.02 %
Cl (Chloride) Max. 20 ppm
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 50 ppm
Na (Sodium) Max. 0.02 %
Sr (Strontium) Max. 0.3 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR