Barium nitrate ≥99.0%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Danger

21740.293EA 160 EUR
21740.293
Barium nitrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Barium nitrate
Formula: Ba(NO₃)₂
MW: 261.34 g/mol
Melting Pt: ∼592 °C
Density: ∼3.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003442
CAS Number: 10022-31-8
EINECS: 233-020-5
UN: 1446
ADR: 5.1 (6.1) II
Merck Index: 13,00986

Order Now

Specification Test Results

Assay Min. 99.0 %
pH (20°C; 5 %) 5.0 - 8.0
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 100 ppm
Not precipitated by H2SO4 (as SO4) Max. 0.05 %
Cl (Chloride) Max. 5 ppm
Ca (Calcium) Max. 20 ppm
Fe (Iron) Max. 2 ppm
K (Potassium) Max. 50 ppm
Na (Sodium) Max. 50 ppm
Sr (Strontium) Max. 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR