Decanting pumps, hand action, OTAL®
Supplier: BURKLE
5005-1200 5005-1800 5005-3000 5005-4000 5005-6000 5005-1000 5005-1500 5005-7000
223-0006EA 85.15 EUR
223-0006 223-1112 223-1115 223-0061 223-0060 223-1130 223-1113 223-1000
Decanting pumps, hand action, OTAL®
Pumps Drum/Transfer Pumps
The telescopic immersion tube adapts itself to the container height on insertion. This ensures good removal of residual liquid and means that the hand pump can be used with different containers.
  • Particularly compact, space-saving hand pump
  • Good pump capacity
  • With universal bungs made of soft PVC, for container openings of 30 to 70 mm Ø
OTAL® hand pump made of PP
OTAL® hand pumps with PP delivery tubes, resistant to weak acids, alkalis and detergents.
OTAL® hand pump made of PVDF
OTAL® hand pump with PVDF delivery tubes, particularly suitable for acidsMore Product Information