DL-Camphor, tablets, TECHNICAL, refined

Supplier: VWR Chemicals
Warning

Synonyms: (±)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, DL-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (±)-1,7,7-Trimethyl-2-norbornanone

22478.260EA 0 EUR
22478.260
DL-Camphor, tablets, TECHNICAL, refined
DL-Camphor
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 204
Melting Pt: ∼180 °C
Density: 0.992 g/cm³ (25 °C)
Flash Pt: 66
Storage Temperature: Ambient
MDL Number: MFCD00074738
CAS Number: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR