Chloroacetyl chloride ≥98%

Supplier: VWR Chemicals
Danger

22678.294EA 0 EUR
22678.294
Chloroacetyl chloride ≥98%
Chloroacetyl chloride
Formula: C₂H₂Cl₂O
MW: 112.94 g/mol
Boiling Pt: 105
Melting Pt: –22.5 °C
Density: 1.426 g/cm³ (20 °C)
Flash Pt: >100 °C
MDL Number: MFCD00000725
CAS Number: 79-04-9
EINECS: 201-171-6
UN: 1752
ADR: 6.1 I
Merck Index: 13,02074

Order Now

Specification Test Results

Assay Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR