Tubing for Vacuum Applications, Tygon® S3™ E-3603
Supplier: Saint Gobain Life Sciences
ACF00027 ACF00029 ACF00038 ACF00053 ACF00007-S ACF00012 ACF00017 ACF00018-S ACF00001 ACF00002 ACF00003 ACF00004 ACF00005 ACF00032 ACF00036 ACF00037 ACF00039 ACF00045 ACF00046 ACF00055 ACF00020-S ACF1S1506 ACF00015 ACF00030 ACF00049 ACF1S1504 ACF00024 ACF00028-S ACF1S1502 ACF1S1517 ACF1S1518 ACF1S1503 ACF00010-S ACF00006 ACF00009 ACF00013 ACF00019 ACF00022 ACF00023 ACF00059 ACF00016 ACF00011-S ACF00014 ACF00034 ACF00025 ACF00033 ACF00054 ACF00057 ACF00050 ACF00064 ACF00069 ACF00070 ACF00073 ACF00074 ACF00078 ACF1S1505
228-1265EA 0 EUR
228-1265 228-1296 228-1267 228-1314 228-1284 228-1288 228-1263 228-1291 228-1278 228-1279 228-1280 228-1281 228-1282 228-1297 228-1299 228-1310 VERNACF00039 228-1312 228-1313 VERNACF00055 228-1264 228-1626 228-1358 228-1266 228-1360 228-1624 VERNACF00024 228-1295 228-1622 228-1628 228-1629 228-1623 228-1286 228-1283 228-1285 228-1289 228-1292 228-1293 228-1294 228-1315 228-1290 228-1287 VERNACF00014 VERNACF00034 228-1433 228-1298 VERNACF00054 VERNACF00057 VERNACF00050 228-1317 228-1318 VERNACF00070 VERNACF00073 VERNACF00074 VERNACF00078 228-1625
Tubing for Vacuum Applications, Tygon® S3™ E-3603
Tubing Transfer Tubing
Non DEHP tubing for laboratory, food and beverage.
  • Outstanding chemical resistance
  • Slips easily over fittings and grips securely for simple laboratory set-ups
  • Contains no BPA or phthalates
  • Lot-to-lot consistency for reproducible results
Tubing handles most inorganic chemicals in the laboratory. It is non oxidising and non contaminating, and less permeable than rubber tubing. The glassy, smooth inner bore helps prevent build up to facilitate cleaning, and coils are marked at 30,4 cm intervals for easy measuring. Concentrated acids and concentratedMore Product Information