Cutter for plastic tubing, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H210100000
229-0013EA 78.7 EUR
229-0013
Cutter for plastic tubing, SP Bel-Art
Cutters
Portable, pocket-sized cutter cuts flexible tubing from 1,6 to 19 mm Ø.
  • Spring-loaded jaw will remain closed for safety when not in use
  • Replaceable, hardened steel blade makes clean and straight cuts