Double spatulas, micro
Supplier: BOCHEM
3340 3341 3342 3343 3344 3350 3351 3345
231-0047EA 4.6 EUR
231-0047 231-0542 231-1353 231-1354 231-1355 231-0543 231-0239 231-0561
Double spatulas, micro
Spatulas
Stainless steel 18/10, anti-magnetic. Double spatulas for a wide range of uses.
  • One end with micro spoon or scoop and one end with micro spatula
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean