Dibutylamine ≥99%, purified
Supplier: VWR Chemicals
Warning
Synonyms: Di-n-butylamine, N-butyl-1-propanamine
23337.293EA 42.1 EUR
23337.293
Dibutylamine ≥99%, purified
Dibutylamine
Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 159
Melting Pt: –62 °C
Density: 0.759 g/cm³ (20 °C)
Flash Pt: 43
Storage Temperature: Ambient
MDL Number: MFCD00009429
CAS Number: 111-92-2
EINECS: 203-921-8
UN: 2248
ADR: 8 (3) II
Merck Index: 13,03058
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0d7d86589224f8565e12dc8e6b30a8de923393f&vid=d2cc023db25785e54555fc105aa8436e65fd9cbc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay Min. 99 %