Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals

Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol

24385.295EA 136 EUR
24385.295 24385.320 24385.460
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 83.5 - 88.0%
Acidity Max. 0.0004 meq/g
Alkalinity Max. 0.0003 meq/g
Density (20/4) 1.220 - 1.230
Esters of fatty acids (as C15H26O6) Max. 0.05 %
Glyceraldehyde Max. 30 ppm
Heavy metals (as Pb) Max. 1 ppm
Ignition residue (SO4) Max. 50 ppm
Cl (Chloride) Max. 1 ppm
NH4 (Ammonium) Max. 5 ppm
SO4 (Sulphate) Max. 5 ppm
Fe (Iron) Max. 1 ppm
Water 13 - 16 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR