2-Methylbutane ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: iso-Pentane

24872.298EA 121 EUR
24872.298 24872.323 24872.260
2-Methylbutane ≥99%, GPR RECTAPUR®
2-Methylbutane
Formula: CH₃CH₂CH(CH₃)₂
Boiling Pt: 28 °C (1013 hPa)
Melting Pt: –160 °C
Density: 0.62 g/cm³ (20 °C)
Flash Pt: –51 °C
Storage Temperature: Ambient
MDL Number: MFCD00009338
CAS Number: 78-78-4
EINECS: 201-142-8
UN: 1265
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=498904a9f8b2618e19d2343aa74e247ec7121f00&vid=c7ed2044255d0d72af23a3fafec4ef6066d25097&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
Boiling point 27.0 - 28.5 °C
Density (20/4) 0.600 - 0.640
Refractive index (20°C) 1.353 - 1.355
Evaporation residue Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR