D-(-)-Mannitol, GPR RECTAPUR®

Supplier: VWR Chemicals

Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

25313.294EA 99.4 EUR
25313.294 25313.363
D-(-)-Mannitol, GPR RECTAPUR®
D(-)-Mannitol
Formula: HOH₂C(CH(OH))₄CH₂OH
Boiling Pt: 290…295 °C (4 hPa)
Melting Pt: 164…169 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064287
CAS Number: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Melting point 165 - 170 °C
Specific optical rotation (8 %; borate) 23 - 24.5 °
Ignition residue (SO4) Max. 0.2 %
Loss on drying (100°C) Max. 0.5 %
Cl (Chloride) Max. 0.02 %
SO4 (Sulphate) Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR