N-Methyl-2-pyrrolidone (NMP) ≥99%, TECHNICAL

Supplier: VWR Chemicals
Danger

25675.294EA 47.7 EUR
25675.294 25675.465
N-Methyl-2-pyrrolidone (NMP) ≥99%, TECHNICAL
N-Methyl-2-pyrrolidone (NMP)
Formula: C₅H₉NO
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
REACH: 01-2119472430-46
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
IR Spectrum Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR