Ammonium sulphate 50% aqueous solution, J.T.Baker®

Supplier: Avantor

Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

2652-38EA 0 EUR
2652-38
Ammonium sulphate 50% aqueous solution, J.T.Baker®
Ammonium sulphate
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
MDL Number: MFCD00003391
CAS Number: 7783-20-2
Merck Index: 13,00559

Order Now

Specification Test Results

Assay ((NH4)2SO4) min. 500 g/l
Density (g/ml) at 20°C 1.239 - 1.253
Escherichia coli (absent) none detected
pH at 20°C 6.8 - 7.2
Salmonella (absent) none detected
Total Aerobic Microbial Count, cfu/ml max. 100
Total Yeast and Mold Count, cfu/ml max. 100
Ammonium Sulfate Solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR