Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent

Supplier: VWR Chemicals
Danger

Synonyms: Azabenzene, Azine

27199.268EA 150 EUR
27199.268 27199.292 27199.326 27199.460
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Pyridine
Formula: C₅H₅N
Boiling Pt: 115 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0.983 g/cm³ (20 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.7 %
IR Spectrum Passes test
Solubility in water Passes test ACS
Acidity or alkalinity Max. 0.2 meq/g
Boiling point 114 - 116 °C
Colouration Max. 10 APHA
Density (20/4) 0.981 - 0.983
Evaporation residue Max. 20 ppm
Substances reducing KMnO4 (as O) Max. 5 ppm
alpha-Picoline Max. 0.2 %
Piperidine Max. 100 ppm
Water Max. 0.1 %
Cl (Chloride) Max. 5 ppm
NH3 (Ammonia) Max. 20 ppm
SO4 (Sulphate) Max. 5 ppm
Al (Aluminium) Max. 0.05 ppm
B (Boron) Max. 0.1 ppm
Ba (Barium) Max. 0.02 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.05 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.2 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.01 ppm
Na (Sodium) Max. 1 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.01 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.05 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR