Sodium acetate ≥98%, GPR RECTAPUR®

Supplier: VWR Chemicals

27650.292EA 48 EUR
27650.292 27650.361
Sodium acetate ≥98%, GPR RECTAPUR®
Sodium acetate
Formula: H₃CCOONa
Boiling Pt: 881.4 °C (1013 hPa)
Melting Pt: 324 °C
Density: 1.528 g/cm³ (20 °C)
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now

Specification Test Results

Assay Min. 98 %
Heavy metals (as Pb) Max. 20 ppm
Cl (Chloride) Max. 0.02 %
SO4 (Sulphate) Max. 0.02 %
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR