Sodium nitrite ≥98%, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Diazotizing salts

27959.298EA 75.5 EUR
27959.298 27959.367 27959.460
Sodium nitrite ≥98%, GPR RECTAPUR®
Sodium nitrite
Formula: NaNO₂
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 284 °C
Density: 2.168 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011118
CAS Number: 7632-00-0
EINECS: 231-555-9
UN: 1500
ADR: 5.1 (6.1),III
Merck Index: 13,08721

Order Now

Specification Test Results

Assay Min. 98 %
Heavy metals (as Pb) Max. 20 ppm
Cl (Chloride) Max. 100 ppm
Fe (Iron) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR