Trisodium phosphate dodecahydrate ≥98%, TECHNICAL

Supplier: VWR Chemicals
Warning

Synonyms: Sodium Phosphate tribasic 12-Hydrate, Sodium phosphate dodecahydrate, Sodium phosphate tribasic dodecahydrate

28032.293EA 30.8 EUR
28032.293 28032.362 28032.460
Trisodium phosphate dodecahydrate ≥98%, TECHNICAL
tri-Sodium phosphate dodecahydrate
Formula: Na₃PO₄·12H₂O
Melting Pt: ∼75 °C
Density: 1.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149198
CAS Number: 10101-89-0
EINECS: 231-509-8
UN: 3262
ADR: 8,III
Merck Index: 12,08808

Order Now

Specification Test Results

Assay Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR