Disodium metasilicate pentahydrate, TECHNICAL

Supplier: VWR Chemicals
Danger

Synonyms: Sodium metasilicate pentahydrate

28092.290EA 49 EUR
28092.290
Disodium metasilicate pentahydrate, TECHNICAL
di-Sodium metasilicate pentahydrate
Formula: Na₂SiO₃·5H₂O
MW: 212.14 g/mol
Melting Pt: 72.2 °C
Density: 1.749 g/cm³ (20 °C)
MDL Number: MFCD00149176
CAS Number: 10213-79-3
EINECS: 229-912-9
UN: 3253
ADR: 8 III
Merck Index: 13,08716

Order Now

Specification Test Results

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR