Sodium sulphate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Synonyms: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt
28111.296EA 62.2 EUR
28111.296 28111.365 28111.260
Sodium sulphate ≥99%, GPR RECTAPUR®
Sodium sulphate
Formula: Na₂SO₄
Boiling Pt: 1429 °C (1013 hPa)
Melting Pt: 888 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay Min. 99 %
Heavy metals (as Pb) Max. 20 ppm
Fe (Iron) Max. 20 ppm
Sodium chloride Max. 0.25 %