Milk powder sampler, Milky
Supplier: BURKLE
5321-0300
300-0022EA 228.55 EUR
300-0022
Milk powder sampler, Milky
Samplers Solid Samplers
Stainless steel construction. Developed for sampling milk powder. The collector is inserted into bags or casks and the sample is removed using a half-shell shaped lance. Insertion depth 385 mm, total length 480 mm, ext.Ø approximately 28 mm.
  • Sterilisable
  • Stainless steel V4A (1.4404)/AISI 316 L
Acc. EN ISO 707 and IDF (Int. Dairy Federation)
Certifications: Acc. EN ISO 707 and IDF (Int. Dairy Federation)