Immersion cylinders for liquid sampling
Supplier: Sampling Systems
5132B-1000 5132B-300 5132B-500 5132S-1000 5132S-300 5132S-500
300-0472EA 558 EUR
300-0472 300-0473 300-0474 300-0475 300-0476 300-0477
Immersion cylinders for liquid sampling
Samplers Fluid Samplers
These immersion cylinders are designed to take liquid samples from open water as well as tanks, drums, wells, and large storage containers. They are available made from either 304 stainless steel or brass, and are available in a range of sizes.
  • Stainless steel or brass construction
  • Available in 300 ml, 500 ml, and 1000 ml sizes
  • Ideal for either mid-point or bottom sampling
The Immersion Cylinder allows for two methods of sampling. When the cylinder reaches the bottom of the tank, it will automatically open. When the cylinder is mid-level, a secondMore Product Information