Specimen bag re-sealable, BiTran®
Supplier: CELLPATH
WAA-0500-12A WAA-0400-12A WAA-0100-12A WAA-0200-12A WAA-0300-12A WAA-0600-12A
720-2350EA 558 EUR
720-2350 720-0019 300-0706 300-0707 300-0708 300-0709
Specimen bag re-sealable, BiTran®
Bags Storage Bags
The BiTran bag system is designed for transporting wet tissue from one place to another and for the short to long–term storage of tissue.
The bags are manufactured from a special barrier film with a unique double zip closure. The barrier film is impermeable to formalin vapour, therefore no hazardous vapour pollutes the laboratory or any other area once the bag is sealed. The double zip seal provides a leak proof closure, which can be easily opened and closed as many times as required.
  • Leak proof
  • Well–proven long–term storage
  • Space saving
  • High visibility