Perchloric acid 0.1 mol/l (0.1 N) in anhydrous acetic acid, AVS TITRINORM Reag. Ph. Eur. volumetric solution

Supplier: VWR Chemicals
Danger

30110.264EA 55 EUR
30110.264 30110.297 30110.320
Perchloric acid 0.1 mol/l (0.1 N) in anhydrous acetic acid, AVS TITRINORM Reag. Ph. Eur. volumetric solution
Perchloric acid
Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: -101
Density: 1.050 g/cm³ (20 °C)
Flash Pt: 40 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 2920
ADR: 8 (3) II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Titer (20°C; real value 0.5 % accuracy) 0.0998 - 0.1002 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR