Trimethylphosphine oxide

Supplier: Thermo Fisher Scientific
Warning

30394.03 30394.06
30394.03EA 170 EUR
30394.03 30394.06
Trimethylphosphine oxide
Trimethylphosphine oxide

CAS No.: 676-96-0 Molecular Formula: (CH3)3P(O) Formula Weight: 92.08 Physical Form: Crystalline Odour: Odourless Melting Point: 140-141° MDL No.: MFCD00013910

Formula: C₃H₉OP
Melting Pt: 140…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00013910
CAS Number: 676-96-0
EINECS: 211-633-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR