VWR® AVS TITRINORM Buffer Solutions, 20 °C

Supplier: VWR Chemicals

32040.298EA 52.4 EUR
32040.298 32096.291 32031.297 32032.291 32033.294 32033.374 32035.291 32036.294 32038.291 32039.261 32039.374 32040.185 32040.378 32041.292 32042.295 32044.268 32044.292 32044.372 32045.262 32045.295 32045.375 32046.265 32046.298 32046.378 32095.184 32095.264 32095.366 32095.377 32096.187 32096.267 32096.360 32096.371 32097.261 98192.290 98213.260 32039.294 32095.297 83601.260 83601.290 83610.600 32032.260 98192.260 83610.610 32097.290 32040.260 32096.400 32044.290 32045.290 32044.292BOM1 32044.292BOM2 32044.372BOM 32095.366BOM 32045.412 32044.418 85680.290 85680.295 85680.260 932045.262 932045.295
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Buffers pH Calibration Buffers

Solutions prepared from AnalaR® NORMAPUR® grade analytical reagents.


  • Reference temperature 20 °C
  • True analytical value on the label to an accuracy of ±0,02 pH units (pH 1,00 to 11,00); ±0,05 for pH 12,00 buffer
  • Traceable to SRM from NIST
  • Batch number and packing date for traceability
  • Certificate of Analysis available at website


Manufactured from analytical grade products presenting high accuracy: 0,02 pH unit from pH 1,00 to 11,00 and 0,05 pH unit for pH 12,0.


Certifications: Certifications: Products with accreditation to ISO 17034 in the table below are produced and certified in accordance with ISO 17034 accredited by BELAC under the certification number 542-RM. All information regarding the use of these CRMs can be found on the Certificate of Analysis.


Ordering information: If significantly large volumes are required please contact your local Avantor sales office for a customised offer.


Packaging: Available in different pack sizes: 100 ml, 500 ml, 1 L, 5 L plastic bottles and bag-in-box (5 and 10 L).

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR