Pharma zone sampler, Novartos Uno
Supplier: BURKLE
5358-0140 5358-1001 5358-0120 5358-1400 5358-0110 5358-0600 5358-0105 5358-0115 5358-0125 5358-0130 5358-0135 5358-0103 5358-0100 5358-0630 5358-1030 5358-1430
300-0124EA 139.4 EUR
300-0124 300-0127 300-0122 300-0128 300-0120 300-0125 300-0119 300-0121 300-0123 300-0201 BURK5358-0135 BURK5358-0103 BURK5358-0100 331-0377 331-0378 331-0379
Pharma zone sampler, Novartos Uno
Samplers Solid Samplers
Novartos Uno sampler with two or three sample chambers, for one sample zone.
  • Novartos Uno for targeted sampling
  • The sample inserts fit precisely and can easily be replaced
  • Inserts are available in volumes from 0 to 4,0 ml (alternative sample volumes in increments of 0,1 ml on request)
  • The insertion depths can easily be read from the laser-engraved depth scale
  • Stainless steel AISI 316L (1.4404)/AISI 316Ti (1.4571)
  • Groove-free and electro-polished
  • 25 mm Ø
Novartos the Pharma zone sampler for sampling according to FDA guidelines for Unit-Dose-Sampling.More Product Information