VWR® Buffer Tablets for Haematology
Supplier: VWR Chemicals
331552SEA 63.4 EUR
331552S 331562U 331932D 331942F
VWR® Buffer Tablets for Haematology
Buffers Microscopy Buffers
These tablets provide a rapid and convenient method of preparing a buffer solution.
Gurr buffer tablets for the preparation of microscopical stain solutions.
One tablet dissolved in distilled water, and made up to 100 ml, produces a solution of the stated pH at 20 °C. The molarity of the buffer solutions is 0,005 M.