VWR® Tap for Drums, Vented
Supplier: VWR Chemicals
332862KEA 76.3 EUR
332862K
VWR® Tap for Drums, Vented
Faucets and Sinks Faucets
PE, white, ³/₄" screw thread.
Supplied with 2" adapter (PE) for 25 and 200 L drums.