Thymol blue 0,04% ethanol, TECHNICAL

Supplier: VWR Chemicals
Danger

Synonyms: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

34300.234EA 59.7 EUR
34300.234 34300.234_T
Thymol blue 0,04% ethanol, TECHNICAL
Thymol blue

Transition range: pH 8,0 - 9,6

Formula: C₂₇H₃₀O₅S
MW: 466.6 g/mol
Melting Pt: 221
Storage Temperature: Ambient
MDL Number: MFCD00005869
CAS Number: 76-61-9
EINECS: 200-973-3
UN: 1170
ADR: 3 II
Merck Index: 12,09541

Order Now

Specification Test Results

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR