Anti-PTPRC Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 30-F11]
Supplier: Tonbo Biosciences
35-0451-U025 35-0451-U500 35-0451-U100
35-0451-U025EA 19.2 EUR
35-0451-U025 35-0451-U500 35-0451-U100
Anti-PTPRC Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 30-F11]
Antibodies
The 30-F11 antibody reacts with mouse CD45, which is one of the most abundant hematopoietic markers and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD4More Product Information