Phosphate, Ion chromatography standard solution, Specpure®, PO₄³⁻ 1000 µg/ml

Supplier: Thermo Fisher Scientific

35548.AE 35548.AP
35548.AEEA 163 EUR
35548.AE 35548.AP
Phosphate, Ion chromatography standard solution, Specpure®, PO₄³⁻ 1000 µg/ml
Standards Anionic Standards Phosphate Standards

Molecular Formula: K₂SO₄ in H₂O
Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00145108

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR