Acetic acid standardised solution 0.1 N

Supplier: Thermo Fisher Scientific

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

35567.K2 35567.M4
35567.K2EA 40.8 EUR
35567.K2 35567.M4
Acetic acid standardised solution 0.1 N
Acetic acid

Molecular Formula: CH3CO2H Formula Weight: 60.05 Physical Form: Liquid MDL No.: MFCD00036152

Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR