Potassium hydrogen phthalate standardised solution 0.05 N in aqueous solution volumetric solution

Supplier: Thermo Fisher Scientific

Synonyms: Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt

35603.AP 35603.K2
35603.APEA 36.6 EUR
35603.AP 35603.K2
Potassium hydrogen phthalate standardised solution 0.05 N in aqueous solution volumetric solution
Potassium hydrogen phthalate

CAS No.: 877-24-7 Molecular Formula: C8H5KO4 Formula Weight: 204.23 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00013070

Formula: HOOCC₆H₄COOK
Storage Temperature: Ambient
MDL Number: MFCD00013070
CAS Number: 877-24-7
EINECS: 212-889-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4652479e822d1591ac7eb5e29f892b8c94588ad4&vid=75071da7f30b07f7bd077699a3b54ec89d80f0b8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR