Sodium hydroxide standardised solution 0.05 N in aqueous solution

Supplier: Thermo Fisher Scientific

Synonyms: Caustic soda

35623.K2 35623.K7 35623.M4
35623.K2EA 38.8 EUR
35623.K2 35623.K7 35623.M4
Sodium hydroxide standardised solution 0.05 N in aqueous solution
Sodium hydroxide

CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00003548

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 III
Merck Index: 14,08627

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR