Nitric acid solution_standardised 0.1 N

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Nitric acid solution

35626.AP 35626.K2
35626.APEA 50.2 EUR
35626.AP 35626.K2
Nitric acid solution_standardised 0.1 N
Nitric acid

CAS No.: 7697-37-2 Molecular Formula: HNO3 Formula Weight: 63.01 Physical Form: Liquid Appearance: Colourless

Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 87
Melting Pt: -42
Density: 1.51
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 3264
ADR: 8 III
Merck Index: 13,06610

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR