Copper ≥99.8% (metals basis), thin foil, Thickness: 0.01 mm (0.0003 in)

Supplier: Thermo Fisher Scientific

35815.FF 35815.FI 35815.GH
35815.FFEA 66.5 EUR
35815.FF 35815.FI 35815.GH
Copper ≥99.8% (metals basis), thin foil, Thickness: 0.01 mm (0.0003 in)
Copper

Purity: 99.8% CAS No.: 7440-50-8 Physical Form: ≈0.06g/25x25mm, not certified pinhole free Odour: Odourless MDL No.: MFCD00010965

Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR