Sodium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Warning

3588-01EA 102.13 EUR
3588-01 3588-05 3588-07
Sodium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Sodium bromide

  • Lot analysis on label

Formula: NaBr
Melting Pt: 753 °C
Density: 3.21 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003475
CAS Number: 7647-15-6
Merck Index: 13,08666

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements ≥99.0%
For Laboratory,Research,or Manufacturing Use ≤0.005%
ACS - Assay (NaBr) ≥99.0 %
ACS - Insoluble Matter ≤0.005 %
ACS - Alkalinity (as Na₂CO₃) ≤0.015 %
ACS - pH of 5% Solution at 25°C 5.0 - 8.8
ACS - Bromate (BrO₃) ≤0.001 %
ACS - Calcium (Ca) ≤0.002 %
ACS - Chloride (Cl) ≤0.2 %
ACS - Sulfate (SO₄) ≤0.002 %
ACS - Barium (Ba) ≤0.002 %
ACS - Magnesium (Mg) ≤0.001 %
ACS - Potassium (K) ≤0.1 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR