di-Ammonium oxalate monohydrate 99.0-101.0% ACS

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Ammonium oxalate monohydrate, Ammonium oxalate-1-hydrate, Oxalic acid diammonium salt monohydrate, Oxalic acid ammonium salt monohydrate, Ammonium ethanedioate monohydrate, Ammonium Oxalate, Monohydrate

36228.22 36228.36 36228.57
36228.22EA 25.9 EUR
36228.22 36228.36 36228.57
di-Ammonium oxalate monohydrate 99.0-101.0% ACS
di-Ammonium oxalate monohydrate

Purity: 99.0-101.0% CAS No.: 6009-70-7 Molecular Formula: (COONH4)2.H2O Formula Weight: 142.11 (124.10anhy) Physical Form: Granular Density: 1.50 Appearance: White Melting Point: 70° dec. Solubility: Soluble in water. Slightly soluble in alcohol Beilstein Reference: 3759830 Merck Reference: 14,537 UN No.: 3288 MDL No.: MFCD00149694


In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metals

Formula: (NH₄)₂C₂O₄·H₂O
Boiling Pt: 70 °C (1013 hPa)
Melting Pt: 70 °C (dec.)
Density: 1.50
Storage Temperature: Ambient
MDL Number: MFCD00149694
CAS Number: 6009-70-7
EINECS: 214-202-3
UN: 3288
ADR: 6.1,III
Merck Index: 14,537

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR