Magnesium oxide substrate 10x10x0.5mm, polished one side 100 orientation

Supplier: Thermo Fisher Scientific

Synonyms: Magnesia usta

36427.KT 36427.KU
36427.KTEA 148 EUR
36427.KT 36427.KU
Magnesium oxide substrate 10x10x0.5mm, polished one side 100 orientation
Magnesium oxide

CAS No.: 1309-48-4 Molecular Formula: MgO MDL No.: MFCD00011109

Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600
Melting Pt: 2800
Density: 3.58 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR