Diethyl ether, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

364331000 364335000 364330010 364330025
364331000.EA 29.8 EUR
364331000. 364335000. 364330010. 364330025.
Diethyl ether, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Diethyl ether
Formula: (CH₃CH₂)₂O
Boiling Pt: 34.6 °C (760 mmHg)
Melting Pt: –116 °C
Density: 0.714 g/cm³
Flash Pt: –45 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance clear colorless liquid
GC ≥99.4%
Water ≤0.005% (Coulometric)
Acidity ≤0.0002 meq/g
Peroxides ≤0.002 % (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR