4-Nitro-3-picoline-N-oxide 98%, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Warning

Synonyms: 4-Nitropicoline N-oxide, 3-Methyl-4-nitropyridine 1-oxide, 3-Methyl-4-nitropyridine-N-oxide, 3-Methyl-4-nitropyridine N-oxide, 4-Nitro-3-picoline N-oxide, 3-Methyl-4-nitropyridine-1-oxide

M-Clarity™ quality level = MQ200

370061-1G 370061-5G
370061-1GEA 128 EUR
370061-1G 370061-5G
4-Nitro-3-picoline-N-oxide 98%, Sigma-Aldrich®
4-Nitro-3-picoline N-oxide

NLO material.

Formula: C₆H₆N₂O₃
Melting Pt: 137.5 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00014626
CAS Number: 1074-98-2
EINECS: 214-050-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=295d4261ff1b92687eee57e12c17849e4f5b801e&vid=1d97a39ea83235463da9c47fa3b187266470e7fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour) Yellow
Appearance (form) Powder or Crystals or Chunk(s)
Purity (HPLC Area %) ≥97.5%
Carbon content 45.6 - 47.9%
Nitrogen content 17.7 - 18.6%
Infrared spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR