Sodium iodide 99,5, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril

3748-07EA 0 EUR
3748-07
Sodium iodide 99,5, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Sodium iodide

Crystals. Lot analysis on label.


  • Lot analysis on label

Formula: NaI
MW: 149.89 g/mol
Boiling Pt: 1300
Melting Pt: 662
Density: 3.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9 III
Merck Index: 13,08705

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements ≥99.5%
For Laboratory,Research,or Manufacturing Use 6.0 - 9.0
ACS - Assay (NaI) ≥99.5 %
ACS - pH of 5% Solution at 25°C 6.0 - 9.0
ACS - Insoluble Matter ≤0.01 %
ACS - Chloride and Bromide (as Cl) ≤0.01 %
ACS - Iodate (IO₃) ≤3 ppm
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Barium (Ba) ≤0.002 %
ACS - Heavy Metals ≤5 ppm
ACS - Iron (Fe) ≤5 ppm
ACS - Calcium (Ca) ≤0.002 %
ACS - Magnesium (Mg) ≤0.001 %
ACS - Potassium (K) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR