Bromocresol green sodium salt 0.04% (w/v) in aqueous solution

Supplier: Thermo Fisher Scientific

38696.AE 38696.AP
38696.AEEA 39.6 EUR
38696.AE 38696.AP
Bromocresol green sodium salt 0.04% (w/v) in aqueous solution
Bromocresol green sodium salt

CAS No.: 62625-32-5 Molecular Formula: C21H13Br4NaO5S Formula Weight: 720.01 Physical Form: Liquid Appearance: Green

Formula: C₂₁H₁₃Br₄NaO₅S
Storage Temperature: Ambient
MDL Number: MFCD00148898
CAS Number: 62625-32-5
EINECS: 263-657-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR