Incubators, IF/IFplus (forced convection)

Supplier: MEMMERT
IF260PLUS IF30 IF30PLUS IF450 IF450PLUS IF55 IF55PLUS IF75 IF750 IF750PLUS IF75PLUS IF110 IF110PLUS IF160 IF160PLUS IF260 IF55M IF55MPLUS IF750M IF750MPLUS IF110M IF110MPLUS IF160M IF75M IF160MPLUS IF260M IF260MPLUS IF75MPLUS IF30MPLUS IF30M IF450M IF450MPLUS
390-0633EA 5119 EUR
390-0633 390-0634 390-0635 390-0636 390-0637 390-0638 390-0639 390-0640 390-0641 390-0642 390-0643 390-0628 390-0629 390-0630 390-0631 390-0632 MEMMIF55M MEMMIF55MPLUS MEMMIF750M MEMMIF750MPLUS MEMMIF110M MEMMIF110MPLUS MEMMIF160M MEMMIF75M MEMMIF160MPLUS MEMMIF260M MEMMIF260MPLUS MEMMIF75MPLUS MEMMIF30MPLUS MEMMIF30M MEMMIF450M MEMMIF450MPLUS
Incubators, IF/IFplus (forced convection)
Incubators Standard Incubators
The range has natural and forced convection models with a temperature range up to 80 °C. There is a choice of two different displays, SingleDISPLAY or TwinDISPLAY depending on the application.

  • SingleDISPLAY models, ideal for standard applications:
  • ControlCOCKPIT parameters: Temperature (Celsius or Fahrenheit), fan speed, exhaust air flap position, program time, time zones and daylight-saving time
  • One PT100 temperature sensor in a four wire circuit and Ethernet interface for exporting the protocol log
  • Double over-temperature protection including visual
More Product Information
Order Now

SPECIFICATIONS

Model IF30 IF450 IF55 IF75 IF750 IF110 IF160 IF260 IF55 IF750 IF110 IF160 IF75 IF260 IF30 IF450
Environmentally Preferable
Temperature fluctuation (time) ≤±0,2 °C @ 37 °C ≤±0,2 °C @ 37 °C ≤±0,2 °C @ 37 °C ≤±0,2 °C @ 37 °C ≤±0,2 °C @ 37 °C ≤±0,2 °C @ 37 °C ≤±0,2 °C @ 37 °C ≤±0,2 °C @ 37 °C ≤±0,2 °C @ 37 °C ≤±0,2 °C @ 37 °C ≤±0,2 °C @ 37 °C ≤±0,2 °C @ 37 °C ≤±0,2 °C @ 37 °C ≤±0,2 °C @ 37 °C ≤±0,2 °C @ 37 °C ≤±0,2 °C @ 37 °C
Temperature variation (spatial) ≤± 0,3 °C @ 37 °C ≤± 0,3 °C @ 37 °C ≤± 0,3 °C @ 37 °C ≤± 0,3 °C @ 37 °C ≤± 0,3 °C @ 37 °C ≤± 0,3 °C @ 37 °C ≤± 0,3 °C @ 37 °C ≤± 0,3 °C @ 37 °C ≤± 0,3 °C @ 37 °C ≤± 0,3 °C @ 37 °C ≤± 0,3 °C @ 37 °C ≤± 0,3 °C @ 37 °C ≤± 0,3 °C @ 37 °C ≤± 0,3 °C @ 37 °C ≤± 0,3 °C @ 37 °C ≤± 0,3 °C @ 37 °C
Capacity 32 L 449 L 53 L 74 L 749 L 108 L 161 L 256 L 53 L 749 L 108 L 161 L 74 L 256 L 32 L 449 L
Convection type Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection
Max. load per shelf 20 kg 30 kg 20 kg 20 kg 30 kg 20 kg 20 kg 20 kg 20 kg 30 kg 20 kg 20 kg 20 kg 20 kg 20 kg 30 kg
Power consumption 800 W 1800 W 1000 W 1250 W 2000 W 1400 W 1600 W 1700 W 1000 W 2000 W 1400 W 1600 W 1250 W 1700 W 800 W 1800 W
Temperature range RT* +10…80 °C RT* +10…80 °C RT* +10…80 °C RT* +10…80 °C RT* +10…80 °C RT* +10…80 °C RT* +10…80 °C RT* +10…80 °C RT* +10…80 °C RT* +10…80 °C RT* +10…80 °C RT* +10…80 °C RT* +10…80 °C RT* +10…80 °C RT* +10…80 °C RT* +10…80 °C
Weight 44 kg 160 kg 55 kg 64 kg 192 kg 72 kg 80 kg 96 kg 55 kg 192 kg 72 kg 80 kg 64 kg 96 kg 44 kg 160 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR