VWR® INCU-Line® Tower Cooled Incubator
Supplier: VWR Collection
390-0963EA 2510 EUR
390-0963
VWR® INCU-Line® Tower Cooled Incubator
Incubators Cooled Incubators
The INCU-Line® Tower is a general purpose, dual-chamber, cooled incubator with natural convection. This digitally cooled incubator with Peltier technology offers an economical, space saving solution for microbiology applications at below room temperature. The built-in Peltier element has low energy consumption (only 35 W at 37 °C), and ensures temperature stability and uniformity over the entire temperature range.
  • Compact design with a footprint of only 0,18 m²
  • Digital PID control with PT100 sensor and LED display (resolution 0,1 °C)
  • Excellent temperature